Ion selective electrode, ammonia
Lieferant: Mettler - Toledo
51341000
662-2809EA 1140 EUR
662-2809
Ion selective electrode, ammonia
Elektroden ISE-Elektroden
This gas-sensing electrode is intended for reliably measuring the ammonia content (NH₃).
  • Modular and versatile
  • Refillable electrolyte
  • Polymer matrix membrane