pH-Elektroden, InLab®
Lieferant: Mettler - Toledo
51343070 51343031 51343050 51343054 51343056
662-2837EA 335 EUR
662-2837 662-2825 662-2826 662-2830 662-2832
pH-Elektroden, InLab®
Elektroden pH-Elektroden
pH electrodes with S7, BNC or MultiPin™ (pH/temperature) connector.
  • Liquid filled electrodes for efficient pH measurements in daily laboratory use of a wide application range
  • ARGENTHAL™ reference system with silver-ion trap so that there is no risk of contamination by sulphides and proteins
  • Conform to RoHS ('Restriction of Hazardous Substances' in electrical and electronic equipment) and WEEE ('Waste Electrical and Electronic Equipment')
  • InLab® Versatile Pro features a replaceable ceramic junction which offers an economic alternative to open or movable sleeve junctions
Junction: Movable glass sleeve
* Integrated temperature probe (NTC30)
** Integrated temperature probe (Pt1000)
Electrolyte: 3 mol/l KCl
Bestellinformation: Supplied with test certificate and with (BNC connection) or without cable (S7 and MultiPin™ connections). Junction tool and set of junctions for replacing the junction of the InLab® Versatile Pro electrode are available separately.