Reference electrode filling solutions
Lieferant: Mettler - Toledo
51343185 51343182 51343184 51343180 51350084 51350076 51350074 51350072 51350082
662-2886EA 12 EUR
662-2886 METT51343182 METT51343184 METT51343180 METT51350084 METT51350076 METT51350074 METT51350072 METT51350082
Reference electrode filling solutions
Elektrolyte Elektrodenfülllösungen
FRISCOLYT-B® (84,4% glycerol, 11% KCl and 4,6% H₂O) is for low temperature samples.