Software for data loggers
Lieferant: Greisinger
601145
662-5511EA 0 EUR
662-5511
Software for data loggers
Software Labor-Software
This windows-software is suitable for EASYLog and T-Logg with logger.
  • Automated retrieval and archiving
  • Data export of series of measurements as a CSV file
  • Print out the data as a table and/or trace
  • Multilingual: German, English, Czech and French
  • Operations: Start, Stop, Cycle time, Read data, Delete data.
Chart display: Various measuring sequences in one diagram, real-time axis, zoom function, measurement points, legend and measurement cursor displayed or hidden, add comments.
System requirements: 1 GHz CPU, 1 GB RAM, 100 MB HDD, Windows 7 SP 1 (32More Product Information