Cleaning and Storage Solutions
Lieferant: HAMILTON BONADUZ
238931
662-9223EA
32.8
EUR
662-9223
Cleaning and Storage Solutions
Elektrolyte
Elektrodenaufbewahrungslösungen
Hamilton storage solution is recommended to increase sensor longevity when not in use. The liquid solution is acid buffered to help with sensor response time after storage as well as prevent any biological growth.