Orion™ Star A122 Conductivity/TDS/°C Meters, Handheld
Lieferant: Thermo Orion
STARA1225 STARA1220
663-0211EA 1050 EUR
663-0211 663-0210
Orion™ Star A122 Conductivity/TDS/°C Meters, Handheld
Elektrochemische Messgeräte Leitfähigkeitsmessgeräte
The Orion™ Star A122 conductivity meter offers basic conductivity analysis in the field. Ideal for taking simple conductivity, TDS and temperature measurements.
  • Perform a one point automatic or manual calibration
  • Auto-ranging conductivity values offer consistent readings across entire measurement range
  • Simplified, easy-to-read display shows main measurement with temperature, stability indicator and electrode condition icon
  • User-friendly operation with basic calibration text prompts
  • Easily recall calibration data for procedure checks
  • Non-volatile memory preserves 50 point internal data log and meter settings, even with loss of power
  • Flexible power options with AA batteries or optional universal power adapter
  • IP 67-rated housing is waterproof for rugged field operation
Users can obtain temperature compensated readings using linear option with 20 or 25 °C reference temperatures. They can quickly navigate setup menus and ensure consistent calibrations using the simple keypad layout and on-screen text prompts. Three year meter warranty.
Standards: TUV 3-in-1 and FCC class A.
Lieferumfang: Meter is supplied with four AA batteries, printed quick start guide and literature CD. Kit contains meter, conductivity probe (K=1,0), 1413 µS conductivity standard, rinse solution pouches, protective armor with electrode holder, hard-sided field case, four AA batteries, printed quick start guide and literature CD.