The Orion™ Star A122 conductivity meter offers basic conductivity analysis in the field. Ideal for taking simple conductivity, TDS and temperature measurements.
- Perform a one point automatic or manual calibration
- Auto-ranging conductivity values offer consistent readings across entire measurement range
- Simplified, easy-to-read display shows main measurement with temperature, stability indicator and electrode condition icon
- User-friendly operation with basic calibration text prompts
- Easily recall calibration data for procedure checks
- Non-volatile memory preserves 50 point internal data log and meter settings, even with loss of power
- Flexible power options with AA batteries or optional universal power adapter
- IP 67-rated housing is waterproof for rugged field operation
Users can obtain temperature compensated readings using linear option with 20 or 25 °C reference temperatures. They can quickly navigate setup menus and ensure consistent calibrations using the simple keypad layout and on-screen text prompts. Three year meter warranty.
Standards: TUV 3-in-1 and FCC class A.
Lieferumfang: Meter is supplied with four AA batteries, printed quick start guide and literature CD. Kit contains meter, conductivity probe (K=1,0), 1413 µS conductivity standard, rinse solution pouches, protective armor with electrode holder, hard-sided field case, four AA batteries, printed quick start guide and literature CD.