Bioprocess pH electrode, InLab® Smart pro-ISM
Lieferant: Mettler - Toledo
30027775
663-0309EA 1050 EUR
663-0309
Bioprocess pH electrode, InLab® Smart pro-ISM
Elektroden pH-Elektroden
This unique digital pH electrode for lab use is really clever: by keeping track of the membrane and reference resistance as well as the average pH and temperature readings, the InLab® Smart Pro-ISM allows for preventative maintenance. Users can effectively save costs, increase uptime respectively.
  • High performance digital sensor with no alkali error
  • Autoclavable
  • Sensor diagnostics functions for increased system uptime
  • Highly stable, maintenance-free SteadyForce™ reference system
  • Integrated temperature probe automatic temperature compensation
  • Only works with SevenExcellence™ and SevenCompact™ (with optional cable)
All functions and benefits that users are used to getting from Intelligent Sensor Management (ISM®), like sensor recognition and the secure handling of the sensor ID and calibration data, calibration history etc. are implemented in this digital sensor too.
Lieferumfang: Supplied with quality certificate.