Dissolved oxygen meter, handheld, Orion Star™ A223
Lieferant: Thermo Orion
STARA2230 STARA2235
664-0147EA 880 EUR
664-0147 664-0148
Dissolved oxygen meter, handheld, Orion Star™ A223
Elektrochemische Messgeräte Sauerstoff-Messgeräte
This portable DO/RDO meter is ideal for a wide range of applications and advanced dissolved oxygen analysis in the field. It can be used with RDO optical or a polarographic DO probes. The meter is suitable for use in the most demanding locations thanks to its IP67-rated housing.
  • Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
  • Measurement stability indicator and selectable read modes – Auto-Read holds a stable reading and Continuous shows changing readings with hold option
  • Data log collects and date/time stamps up to 1000 measurement sets
  • Non volatile meter memory ensures your data and settings are protected, even with loss of power
  • Orion Star Com software facilitates data transfer to a computer via USB or RS232 connections
  • Flexible power options with AA batteries or optional universal power adapter
Calibrate dissolved oxygen using water-saturated air, air-saturated water, custom value (Winkler titration) and zero point options. Automatically compensate dissolved oxygen readings using ATC temperature, manual salinity input and built-in barometer for barometric pressure readings.
Standards: CE, TUV 3-in-1 and FCC Class A
Lieferumfang: Supplied with four AA batteries, literature CD, printed quick start guide, computer interface cable, meter test certificate and 3-year replacement warranty.
The kit additionally includes: 087010MD RDO optical DO sensor with 3 m cable, calibration sleeve and guard, Orion Star A series portable meter armor and Orion Star A series hard-sided field case.