Oxygen Sensor Agile, Memosens COL37E
Lieferant: Endress+Hauser
COL37E-1020/0
664-0242EA 1250 EUR
664-0242
Oxygen Sensor Agile, Memosens COL37E
Elektroden Sauerstoff-Elektroden
Optical oxygen sensor with stainless steel for laboratory measurements and random sampling in the field. Connectable via inductive, digital connection with Memosens technology.
  • C-shaped optical cap for stable measurements without interference from air bubbles
  • Excellent performance and low maintenance due to optical oxygen measuring principle
The optical Memosens 2.0 oxygen sensor COL37E can be used in all environments and enables a quick check of oxygen levels in aeration tanks, fish farms and surface water.
Thanks to Memosens technology, the sensors provide reliable, highly accurate measurements with a fast response time.
The inductive Memosens coupling between the corresponding Memosens sensor and the mobile CML18 handheld device is insensitive to moisture and corrosion
The Memosens technology is based on a true plug-and-play principle. Just connect the sensor and you are ready to start your measurements. Measured values, calibration data and history are stored directly in the sensor. This means status values can be read out and a sensor check is also possible. An excellent feature to boost your GLP.
All sensors are supplied pre-calibrated. Thus, they are directly ready for use for your random sample measurement when the corresponding sensor is connected to the associated mobile handheld device Liquiline Mobile CML18.
Standards: EC_00828_03.22; EU_01001_01.22; UK_00483_01.22. Nonhazardous area approval. CE + UKCA marking.
Zubehörinformationen: The COL37E maintenance kit contains optical cap, O-rings, and further material for maintenance of the Memosens COL37E oxygen sensor.