pH/mV/ISE/°C meters, bench top, Orion Star™ A214
Lieferant: Thermo Orion
STARA2140 STARA2145 STARA2146 STARA2147 STARA2148
665-0269EA 1390 EUR
665-0269 665-0270 665-0477 665-0478 665-0479
pH/mV/ISE/°C meters, bench top, Orion Star™ A214
Elektrochemische Messgeräte pH- und Multi-Parameter-Messgeräte
These bench top meters record accurate and reliable pH, ion concentration, mV, ORP and temperature measurements. They are ideal for a wide range of applications and advanced pH or ion analysis in the lab.
  • Up to a five point pH calibration using automatic buffer recognition of NIST or DIN buffer groups
  • Up to a five point ISE (Ion Selective Electrode) calibration with selectable units of ppm, M, mg/L, %, ppb or none
  • Advanced ISE features include non-linear selectable auto-blank and low range stability
  • Measurement stability indicator and selectable read modes – Auto-Read holds a stable reading, Continuous shows changing readings with hold option and Timed logs data at specific intervals
  • Data log collects and date/time stamps up to 2000 measurement sets with optional sample ID and user ID
  • Calibration log stores ten most recent calibrations
  • Orion Star Com software facilitates data transfer to a computer via USB or RS232 connections
  • Comprehensive with menu-specific function keys and dual purpose scroll/shortcut keys
Can be used with a wide variety of Orion ion selective electrodes including electrodes that measure ammonia, ammonium, bromide, cadmium, calcium, carbon dioxide, chloride, chlorine, cupric, cyanide, fluoride, fluoroborate, iodide, lead, nitrate, potassium, silver, sodium, sulfate, surfactant and thiocyanate concentrations.
Use the included electrode stand to easily place sensors in samples and simplify storage to minimise breakage. Eliminate the need for magnetic stir bars using the meter-controlled stirrer probe (catalog number 096019) to uniformly mix solutions.
Standards: TUV 3-in-1 and FCC Class A.
Lieferumfang: Supplied with attached electrode stand and holder, universal power adapter, printed quick start guide and literature CD and USB computer cable.
The first kit additionally includes 8102BNUWP Orion ROSS Ultra pH electrode, 927007MD Orion stainless steel ATC probe, 096019 Orion Star stirrer probe and 810199 ROSS pH buffer and solution kit.
The second kit additionally includes 8102BNUWP Orion ROSS Ultra pH electrode, 9512HPBNWP Orion high-performance, ammonia ISE, 927007MD Orion stainless steel ATC probe, 096019 Orion Star stirrer probe, 951007 Ammonia 1000 ppm standard, 951210 Low-level ammonia ISA, 951213 Ammonia ISE storage solution.
The third kit additionally includes 8102BNUWP Orion ROSS Ultra pH electrode, 9609BNWP Orion fluoride ISE, 927007MD Orion stainless steel ATC probe, 096019 Orion Star stirrer probe, 040906, 040907 and 040908 Orion fluoride, standards; 1, 2 and 10 ppm with TISAB II, 940909 Orion TISABII solution.
The fourth kit additionally includes 8102BNUWP Orion ROSS Ultra pH electrode, 8611BNWP Orion ROSS sodium ISE and sodium solutions, 927007MD Orion stainless steel ATC probe, 096019 Orion Star stirrer probe.