Uridin-5'-triphosphorsäure Trinatriumsalz (UTP-Na3), Millipore®
Lieferant: Merck Millipore Calbiochem
Synonyme:
Uridine-5'-triphosphoric acid trisodium salt, Uridine triphosphate trisodium salt
6701-50
6701-50EA
84.9
EUR
6701-50
Uridin-5'-triphosphorsäure Trinatriumsalz (UTP-Na3), Millipore®
Uridin-5'-triphosphorsäure Trinatriumsalz (UTP-Na3)
Formel:
C₉H₁₂N₂Na₃O₁₅P₃ Molecular Weight: 550,09 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00044310 CAS-Nummer: 19817-92-6 EINECS: 243-347-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...