Acetonitril, ChromAR® für die HPLC, Macron Fine Chemicals™
Lieferant: Avantor Performance Materials
6711-25EA
144.24
EUR
6711-25
Acetonitril, ChromAR® für die HPLC, Macron Fine Chemicals™
Acetonitril
Formel:
H₃CCN |
MDL:
MFCD00001878 CAS-Nummer: 75-05-8 UN: 1648 ADR: 3,II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...