Kaliumnitrat 99,0-100,5%, feine Kristalle, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials
Achtung

Synonyme: Kalisalpeter

6715-20EA 0 EUR
6715-20
Kaliumnitrat 99,0-100,5%, feine Kristalle, Macron Fine Chemicals™
Kaliumnitrat
Formel: KNO₃
Schmelzpunkt: 334 °C
Dichte: 2,109 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011409
CAS-Nummer: 7757-79-1
EINECS: 231-818-8
UN: 1486
ADR: 5.1,III
REACH: 01-2119488224-35
Merck Index: 13,07733

Bestellen

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use 99.0 - 100.5 %
Assay (KNO₃) 99.0 - 100.5 %
Solubility Passes Test
Loss on Drying at 105°C ≤1.0 %
Reaction Passes Test
Identification A Passes Test
Identification B Passes Test
Identification for Sodium Passes Test
Sodium (Na), For Information Only Passes Test
Chlorate Passes Test
Chloride (Cl) ≤0.058 %
Arsenic (As) ≤0.0003 %
Heavy Metals (as Pb) ≤0.0020 %
Lead (Pb) ≤0.0010 %
Appearance (white to off-white crystals)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR