n-Pentan 99, ChromAR® HPLC, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials
Gefahr

Synonyme: Diethylmethan, Pentan, 1,3-Dimethylpropan

6721-25EA 253.23 EUR
6721-25
n-Pentan 99, ChromAR® HPLC, Macron Fine Chemicals™
n-Pentan

Suitable for Liquid Chromatography and UV-Spectrophotometry

Formel: H₃C(CH₂)₃CH₃
Molecular Weight: 72.15 g/mol
Siedepunkt: 36.1 °C
Schmelzpunkt: -130
Dichte: 0.63
Flammpunkt: –48 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00009498
CAS-Nummer: 109-66-0
UN: 1486
ADR: 3 II
Merck Index: 13,07193
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba2c9c81a4b1f44769627a00ba49b6cc8e63132&vid=f6831eb15ab1602d8c752468dc3ee31cc4ee61c0&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (GC) min. 99 %
Residue after Evaporation max. 0.0005 %
Titrable Acid (meq/g) max. 0.0005
Water (KF) max. 0.01 %
Ultraviolet Absorbance (1.00-cm cell vs water):
at 210 nm max. 0.4
at 220 nm max. 0.05
at 230 nm max. 0.02
at 240 nm max. 0.01
at 250 nm max. 0.004


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR