Supelco® TraceCERT® Organic Reference Standard, Bis(2-ethylhexyl) phthalate
Lieferant: Merck
67261-100MG
67261-100MGEA
83.9
EUR
67261-100MG
Supelco® TraceCERT® Organic Reference Standard, Bis(2-ethylhexyl) phthalate
Standards
Organische Standards
- GHS signal: GHS08
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Bitte beachten P201 - P202 - P280 - P308 + P313 - P405 - P501