Diethylether, ChromAR® für die HPLC, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials

6727-25EA 82.2 EUR
6727-25
Diethylether, ChromAR® für die HPLC, Macron Fine Chemicals™
Diethylether
Formel: (CH₃CH₂)₂O
MDL: MFCD00011646
CAS-Nummer: 60-29-7
UN: 1155
ADR: 3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=3dd6c545e58e36e1004e828026523fb0e3bcbbcd&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR