Vacuolin-1 ≥95% (durch HPLC), Calbiochem®, Sigma-Aldrich®
Lieferant: Merck Millipore Calbiochem
Synonyme:
4-(2-(3-Iodobenzylidene)hydrazinyl)-6-morpholino-N,N-diphenyl-1,3,5-triazin-2-amine, 3-Iodobenzaldehyde[4-(diphenylamino)-6-(4-morpholinyl)-1,3,5-triazin-2-yl]hydrazone
Verpackt unter Schutzgas
A cell-permeable triazine based compound that inhibits Ca²⁺-dependent fusion of lysosomes with the plasma membrane and the release of lysosomal content, without affecting enlargeosomes or other membrane-bound organelles.
It does not disturb the organisation of the actin or tubulin cytoskeletal network, nor does it interfere with membrane traffic along the constitutive pathway from the ER to the plasma membrane.
Bitte beachten Protect from light.
Formel:
C₂₆H₂₄IN₇O Molecular Weight: 577,42 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00321410 CAS-Nummer: 351986-85-1 |
Spezifikation Testergebnisse
Purity (by HPLC) | ≥95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...