Gold Standard for ICP-MS, TraceCERT®, Supelco®
Lieferant: Merck
67363-100ML
67363-100MLEA 319 EUR
67363-100ML
Gold Standard for ICP-MS, TraceCERT®, Supelco®
Standards Kationischer Standards Gold-Standards
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034