Sudan IV, Supelco®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ100
67386-25MG
67386-25MGEA
149
EUR
67386-25MG
Sudan IV, Supelco®
Sudan IV
Formel:
C₂₄H₂₀N₄O Schmelzpunkt: 188 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003893 CAS-Nummer: 85-83-6 EINECS: 201-635-8 Merck Index: 12,08538 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...