Supelco® TraceCERT® Organic Reference Standard, ʟ-Lysine monohydrochloride
Lieferant: Merck
67448-100MG
67448-100MGEA 86.4 EUR
67448-100MG
Supelco® TraceCERT® Organic Reference Standard, ʟ-Lysine monohydrochloride
Standards Organische Standards
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43fe57832e4fbf300880526adfc953deab214a27&vid=43fe57832e4fbf300880526adfc953deab214a27&prid=web&clid=SAPDAM