Sodium-2-propylvalerate ≥98% (durch HPLC), Millipore®

Lieferant: Merck Millipore Calbiochem

Synonyme: Sodium Valproate, 2-Propylvaleric Acid Sodium Salt, 2-Propylpentanoic Acid Sodium Salt, Sodium 2-Propylpentanoate, Valproic Acid Sodium Salt

Verpackt unter Schutzgas

676380-5
676380-5EA 101 EUR
676380-5
Sodium-2-propylvalerate ≥98% (durch HPLC), Millipore®
Sodium-2-propylvalerate

Valproic acid is a cell-permeable, short-chained fatty acid that inhibits histone deacetylase.


  • White solid


It induces differentiation and inhibits proliferation of cell lines derived from human malignant gliomas. At therapeutic levels (0,35 - 1,04 mM), causes inositol depletion, inhibits both GSK-3α and -3β, activates ERK pathway and produces neurotropic effects. It also been used as an anti-epileptic agent and reported to stimulate peroxisome proliferator-activated receptor (PPAR) activity. It displays a potent teratogenic activity in humans and rodent models.

Formel: C₈H₁₅O₂Na
Molecular Weight: 166,20 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00078604
CAS-Nummer: 1069-66-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR