Dichlormethan, AR®, Laborreagens, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials
Achtung

6779-45EA 407.49 EUR
6779-45 6779-25
Dichlormethan, AR®, Laborreagens, Macron Fine Chemicals™
Dichlormethan
Formel: CH₂Cl₂
Siedepunkt: 39,8 °C (1013 hPa)
Lagertemperatur: Raumtemperatur
MDL: MFCD00000881
CAS-Nummer: 75-09-2
UN: 1593
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR