Xylol (Isomerengemisch) ≥98,0%, AR®, Macron Fine Chemicals™
Lieferant: Avantor Performance Materials
Synonyme:
Dimethylbenzol
6781-25EA
137.68
EUR
6781-25
Xylol (Isomerengemisch) ≥98,0%, AR®, Macron Fine Chemicals™
Xylol (Isomerengemisch)
Formel:
C₆H₄(CH₃)₂ Molecular Weight: 106.17 g/mol Siedepunkt: 140 Flammpunkt: 25 |
MDL:
MFCD00077264 CAS-Nummer: 1330-20-7 UN: 1307 ADR: 3 III Merck Index: 13,10136 |
|
Spezifikation Testergebnisse
Assay (GC) | Min. 98,0% |
Water (KF) | Max. 0,05% |
Titrable acid (meq/g) | Max. 0,0005 |
Residue after evaporation | Max. 0,002% |
Substances darkened by H₂SO₄ | Passes test |
Sulphur compounds | Max. 0,001% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...