Cyclohexanon, AR®, Laborreagens, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials
Gefahr

6787-25EA 116.15 EUR
6787-25
Cyclohexanon, AR®, Laborreagens, Macron Fine Chemicals™
Cyclohexanone
Formel: C₆H₁₀(=O)
Siedepunkt: 154,3…155.6 °C
MDL: MFCD00001625
CAS-Nummer: 108-94-1
UN: 1145
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8701dac7edb70a452483cd4f1bd716ca29ba9aa&vid=0360daffe7606174162954ba58a6ba2b78e28dfb&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR