ω,4-Dibromacetophenon ≥99,0% (durch HPLC), LiChropur™ Derivatisierungsreagenz, für die HPLC, Supelco®

Lieferant: Merck
Gefahr

Synonyme: 2-Brom-1-(4-bromphenyl)-1-ethanon, Tau Protein Kinase I Inhibitor, 2,4'-Dibromacetophenon, Glycogen Synthase Kinase-3β Inhibitor VII, GSK-3β Inhibitor VII, 4-Bromphenacylbromid, TPK I Inhibitor, α,4-Dibromacetophenon

68082-5G
68082-5GEA 56 EUR
68082-5G
ω,4-Dibromacetophenon ≥99,0% (durch HPLC), LiChropur™ Derivatisierungsreagenz, für die HPLC, Supelco®
ω,4-Dibromacetophenon

Derivatisation reagent for HPLC separation of free fatty acids (FFAs) in human plasma, for liquid chromatography-UV-tandem mass spectrometric analysis of perfluorinated carboxylic acids.

Formel: C₈H₆Br₂O
Molecular Weight: 277,94 g/mol
MDL: MFCD00000200
CAS-Nummer: 99-73-0
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de051f08e3c7a4bea858f19678ade95b8e8e65b9&vid=de051f08e3c7a4bea858f19678ade95b8e8e65b9&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR