Kaliumselenocyanat 98,5

Lieferant: Thermo Fisher Scientific
Gefahr

68107.09 68107.18
68107.09EA 96.9 EUR
68107.09 68107.18
Kaliumselenocyanat 98,5
Potassium selenocyanate

Purity: 98.5% CAS No.: 3425-46-5 Molecular Formula: KSeCN Formula Weight: 144.08 Physical Form: Crystalline Density: 2.347 Appearance: White to off-white Odour: Odourless Melting Point: 162-164° UN No.: 3283 MDL No.: MFCD00011366

Formel: CKNSe
Molecular Weight: 144.08 g/mol
Schmelzpunkt: 162…164 °C
Dichte: 2.347
Lagertemperatur: Raumtemperatur
MDL: MFCD00011366
CAS-Nummer: 3425-46-5
EINECS: 222-320-1
UN: 3283
ADR: 6.1 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR