Seleninyldichlorid 99 Metals basis
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Selen(IV)dichloridoxid, Selen(IV)oxydichlorid
68108.AB
68108.AC
68108.ABEA
362
EUR
68108.AB
68108.AC
Seleninyldichlorid 99 Metals basis
Seleninyldichlorid
Purity: 99%
CAS No.: 7791-23-3
Molecular Formula: SeOCl2
Formula Weight: 165.87
Physical Form: Liquid
Density: 2.42
Refractive Index: 1.651
Sensitivity: Moisture Sensitive
Appearance: Yellow
Melting Point: 8.5°
Boiling Point: 176.4°
Solubility: Decomposes in water forming HCl and H2SeO3
Merck Reference: 14,8435
UN No.: 2879
MDL No.: MFCD00016327
Formel:
Cl₂OSe Molecular Weight: 165.86 g/mol Siedepunkt: 176.4 °C Schmelzpunkt: 8.5 Dichte: 2.42 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016327 CAS-Nummer: 7791-23-3 EINECS: 232-244-0 UN: 2879 ADR: 8 I Merck Index: 14,8435 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...