Wortmannin, Millipore®

Lieferant: Merck Millipore Calbiochem

Synonyme: (1S,6bR,9aS,11R,11bR)-1-(Methoxymethyl)-9a,11b-dimethyl-3,6,9-trioxo-1,6,6b,7,8,9,9a,10,11,11b-decahydro-3H-furo[4,3,2-de]indeno[4,5-h]isochromen-11-yl acetate

681675-1
681675-1EA 145 EUR
681675-1
Wortmannin, Millipore®
Wortmannin

Wortmannin is a cell-permeable, fungal metabolite that acts as a potent, selective and irreversible inhibitor of phosphatidylinositol 3-kinase in purified preparations and cytosolic fractions (IC50 = 5 nm).


  • Blocks the catalytic activity of PI 3-kinase without affecting the upstream signaling events
  • Inhibits MAP kinase activation induced by platelet activating factor in guinea pig neutrophils (200 - 300 nm)
  • A 10 mm (1 mg/233 µl) solution of Wortmannin in DMSO is also available


Preincubation of fibroblasts with wortmannin abolishes PDGF-mediated Ins(3,4,5)P3 formation (IC50 = 5 nm). Also blocks the metabolic effects of insulin in isolated rat adipocytes without affecting the insulin receptor tyrosine kinase activity. Also inhibits other kinases such as myosin light chain kinase (IC50 = 200 nm) and PI 4-kinase at concentrations higher than that required for inhibition of PI 3-kinase. Also blocks phospholipase D.

Formel: C₂₃H₂₄O₈
Molecular Weight: 428,44 g/mol
MDL: MFCD00133927
CAS-Nummer: 19545-26-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR