Kaliumchlorid 99,0-100,5% USP, FCC, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials
Achtung

Synonyme: Potassiumchloride

6838-20EA 0 EUR
6838-20
Kaliumchlorid 99,0-100,5% USP, FCC, Macron Fine Chemicals™
Potassium chloride
Formel: KCl
Molecular Weight: 74,55 g/mol
Siedepunkt: 1420 °C (1013 hPa)
Schmelzpunkt: 773 °C
Dichte: 1,98 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011360
CAS-Nummer: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621

Bestellen

Spezifikation Testergebnisse

Meets U.S.P Requirements
Meets F.C.C. Requirements
GMP Manufactured Product
Food GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
USP - Assay (KCl) (by Ag titrn) 99.0 - 100.5 %
USP - Acidity or Alkalinity (no pink color then pink) Passes Test
USP - Bromide (Br)(USP) ≤0.1 %
USP - Calcium and Magnesium Passes Test
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Iodide (I)(USP) ≤0.005 %
USP - Loss on Drying at 105°C ≤1.0 %
USP - Sodium (Na) Passes Test
FCC - Assay (KCl) (by Ag titrn) 99.0 - 100.5 %
FCC - Acidity or Alkalinity (no pink color then pink) Passes Test
FCC - Bromide and/or Iodide (FCC) Passes Test
FCC - Heavy Metals (as Pb)(FCC) ≤5 mg/kg
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Loss on Drying at 105°C ≤1.0 %
FCC - Sodium (Na) Passes Test
Solubility (10 in 50) Passes Test
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR