Iron Standard for ICP-MS, TraceCERT®, Supelco®
Lieferant: Merck
68436-100ML
68436-100MLEA 255 EUR
68436-100ML
Iron Standard for ICP-MS, TraceCERT®, Supelco®
Standards Kationischer Standards Eisen-Standards
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034