Sudan III, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

68562-25MG
68562-25MGEA 149 EUR
68562-25MG
Sudan III, Supelco®
Sudan III
Formel: C₂₂H₁₆N₄O
MDL: MFCD00003905
CAS-Nummer: 85-86-9
EINECS: 201-638-4
Merck Index: 12,09054
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=013f0717fb66df2a30e12f94501ead944f3b34c9&vid=89ea19842f1772a57b0e76e133b22e18af9a8642&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR