Kaliumchromat ≥99,5%, Kristalle, AR® ACS, Macron Fine Chemicals™
Lieferant: Avantor Performance Materials
|
Gefahr
|
6870-20EA
0
EUR
6870-20
Kaliumchromat ≥99,5%, Kristalle, AR® ACS, Macron Fine Chemicals™
Potassium chromate
Formel:
K₂CrO₄ Molecular Weight: 194,19 g/mol Schmelzpunkt: 985 °C |
MDL:
MFCD00011368 CAS-Nummer: 7789-00-6 UN: 3288 ADR: 6.1,III |
Spezifikation Testergebnisse
Meets ACS Reagent Chemical Requirements | — |
For Laboratory,Research,or Manufacturing Use |
|
ACS - Assay (K₂CrO₄) | ≥ 99.5 % |
ACS - Calcium (Ca) | ≤ 0.005 % |
ACS - Chloride (Cl) | ≤ 0.005 % |
ACS - Insoluble Matter | ≤ 0.005 % |
ACS - pH of 5% Solution at 25°C | 8.6 - 9.8 |
ACS - Sodium (Na) | ≤ 0.02 % |
ACS - Sulfate (SO₄) | ≤ 0.03 % |
Appearance (lemon yellow granular crystals) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...