Molybdenum standard for ICP, TraceCERT®, Supelco®
Lieferant: Merck
68780-100ML
68780-100MLEA
111
EUR
68780-100ML
Molybdenum standard for ICP, TraceCERT®, Supelco®
Standards
Kationischer Standards
Molybdän-Standards
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034