4-Hydroxybenzoesäureethylester 98,0-102,0%, GenAR® NF, Macron Fine Chemicals™
Lieferant: Avantor Performance Materials
Synonyme:
p-Hydroxybenzoesäureethylester, 4-Hydroxybenzoesäureethylester, Ethylparaben
6882-18EA
1387.35
EUR
6882-18
4-Hydroxybenzoesäureethylester 98,0-102,0%, GenAR® NF, Macron Fine Chemicals™
4-Hydroxybenzoesäureethylester
Only Class 3 Solvents are likely to be present. Residual concentration of all Class 3 Solvents is <0,5%.
Formel:
C₉H₁₀O₃ Molecular Weight: 166,18 g/mol |
MDL:
MFCD00002353 CAS-Nummer: 120-47-8 |
Spezifikation Testergebnisse
Meets J.P. Chemical Specifications | |
GMP Manufactured Product | |
Meets N.F. Requirements | |
Meets B.P. Chemical Specifications | |
Meets E.P. Chemical Specifications | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | 98.0 - 102.0% |
NF - Assay (C₉H₁₀O₃) | 98.0 - 102.0 % |
NF - Appearance (white, crystalline powder) | Passes Test |
NF - Color of Solution | Passes Test |
NF - Identification A | Passes Test |
NF - Identification B | Passes Test |
NF - Acidity | Passes Test |
NF - Related Substances - p-Hydroxybenzoic Acid | ≤0.5 % |
NF - Related Substances - Unspecified Impurities (each) | ≤0.5 % |
NF - Related Substances - Total Impurities | ≤1.0 % |
NF - Residue on Ignition | ≤0.1 % |
EP/BP - Assay (C₉H₁₀O₃) | 98.0 - 102.0 % |
EP/BP - Appearance of Solution | Passes Test |
EP/BP - Identification A | Passes Test |
EP/BP - Identification B | Passes Test |
EP/BP - Acidity | Passes Test |
EP/BP - Related Substances - Impurity A | ≤0.5 % |
EP/BP - Related Substances - Unspecified Impurities (each) | ≤0.5 % |
EP/BP - Related Substances - Total Impurities | ≤1.0 % |
EP/BP - Sulfated Ash | ≤0.1 % |
JP - Acidity | Passes Test |
JP - Assay | 98.0 - 102.0 % |
JP - Clarity and Color of Solution | Passes Test |
JP - Identification | Passes Test |
JP - Melting Point | 115 - 118 °C |
JP - Heavy Metals | ≤20 ppm |
JP-RelatedSubstances- p-Hydroxybenzoic Acid | ≤0.5 % |
JP-Related Substances-Unspecified Impurities (each) | ≤0.5 % |
JP-Related Substances-Total Impurities | ≤1.0 % |
JP - Residue on Ignition | ≤0.10 % |
Only Class 3 Solvents are likely to be present. Residual concentration of all Class 3 Solvents is <0.5%. | |
Elemental Impurities (USP 232, EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...