Schwefelsäure 96% ≥96%, ULTREX® II, ultrarein, J.T.Baker®

Lieferant: Avantor
Gefahr

6902.0500EA 778.71 EUR
6902.0500
Schwefelsäure 96% ≥96%, ULTREX® II, ultrarein, J.T.Baker®
Schwefelsäure
Formel: H₂SO₄
Siedepunkt: ∼335 °C (1013 hPa)
Schmelzpunkt: 3 °C
Dichte: 1,84 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064589
CAS-Nummer: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8,II
Merck Index: 14,08974

Bestellen

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use
Assay (H₂SO₄)(w/w) 93 - 98 %
Trace Impurities - Aluminum (Al) ≤ 50 ppt
Trace Impurities - Antimony (Sb) ≤ 50 ppt
Trace Impurities - Arsenic (As) ≤ 500 ppt
Trace Impurities - Barium (Ba) ≤ 10 ppt
Trace Impurities - Beryllium (Be) ≤ 10 ppt
Trace Impurities - Bismuth (Bi) ≤ 10 ppt
Trace Impurities - Cadmium (Cd) ≤ 10 ppt
Trace Impurities - Calcium (Ca) ≤ 50 ppt
Trace Impurities - Cerium (Ce) ≤ 10 ppt
Trace Impurities - Cesium (Cs) ≤ 10 ppt
Trace Impurities - Chromium (Cr) ≤ 10 ppt
Trace Impurities - Cobalt (Co) ≤ 10 ppt
Trace Impurities - Copper (Cu) ≤ 10 ppt
Trace Impurities - Dysprosium (Dy) ≤ 10 ppt
Trace Impurities - Erbium (Er) ≤ 10 ppt
Trace Impurities - Europium (Eu) ≤ 10 ppt
Trace Impurities - Gadolinium (Gd) ≤ 10 ppt
Trace Impurities - Gallium (Ga) ≤ 10 ppt
Trace Impurities - Germanium (Ge) ≤ 100 ppt
Trace Impurities - Hafnium (Hf) ≤ 10 ppt
Trace Impurities - Holmium (Ho) ≤ 10 ppt
Trace Impurities - Indium (In) ≤ 10 ppt
Trace Impurities - Iron (Fe) ≤ 50 ppt
Trace Impurities - Lanthanum (La) ≤ 10 ppt
Trace Impurities - Lead (Pb) ≤ 10 ppt
Trace Impurities - Lithium (Li) ≤ 10 ppt
Trace Impurities - Lutetium (Lu) ≤ 10 ppt
Trace Impurities - Magnesium (Mg) ≤ 50 ppt
Trace Impurities - Manganese (Mn) ≤ 10 ppt
Trace Impurities - Mercury (Hg) ≤ 100 ppt
Trace Impurities - Molybdenum (Mo) ≤ 10 ppt
Trace Impurities - Neodymium (Nd) ≤ 10 ppt
Trace Impurities - Nickel (Ni) ≤ 50 ppt
Trace Impurities - Niobium (Nb) ≤ 10 ppt
Trace Impurities - Palladium (Pd), For Information Only
Trace Impurities - Platinum (Pt), For Information Only
Trace Impurities - Potassium (K) ≤ 50 ppt
Trace Impurities - Praseodymium (Pr) ≤ 10 ppt
Trace Impurities - Rhodium (Rh) ≤ 50 ppt
Trace Impurities - Rubidium (Rb) ≤ 10 ppt
Trace Impurities - Samarium (Sm) ≤ 10 ppt
Trace Impurities - Scandium (Sc) ≤ 10 ppt
Trace Impurities - Selenium (Se) ≤ 500 ppt
Trace Impurities - Silver (Ag) ≤ 50 ppt
Trace Impurities - Sodium (Na) ≤ 50 ppt
Trace Impurities - Strontium (Sr) ≤ 10 ppt
Trace Impurities - Tantalum (Ta), For Information Only
Trace Impurities - Tellurium (Te) ≤ 50 ppt
Trace Impurities - Terbium (Tb) ≤ 10 ppt
Trace Impurities - Thallium (Tl) ≤ 10 ppt
Trace Impurities - Thorium (Th) ≤ 10 ppt
Trace Impurities - Thulium (Tm) ≤ 10 ppt
Trace Impurities - Tin (Sn) ≤ 50 ppt
Trace Impurities - Titanium (Ti) ≤ 50 ppt
Trace Impurities - Tungsten (W) ≤ 10 ppt
Trace Impurities - Uranium (U) ≤ 10 ppt
Trace Impurities - Vanadium (V) ≤ 10 ppt
Trace Impurities - Ytterbium (Yb) ≤ 10 ppt
Trace Impurities - Yttrium (Y) ≤ 10 ppt
Trace Impurities - Zinc (Zn) ≤ 50 ppt
Trace Impurities - Zirconium (Zr) ≤ 10 ppt
Certificate Provided Reports Actual Lot Analysis


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR