4-Phenyl-α-cyanocinnamamide ≥98,5% (durch HPLC) Matrixsubstanz für MALDI-MS, Supelco®
Lieferant: Merck
69028-100MG
69028-100MGEA
161
EUR
69028-100MG
4-Phenyl-α-cyanocinnamamide ≥98,5% (durch HPLC) Matrixsubstanz für MALDI-MS, Supelco®
4-Phenyl-α-cyanocinnamamide
Formel:
C₁₆H₁₂N₂O Molecular Weight: 248,28 g/mol |
CAS-Nummer:
116374-04-0 |
Spezifikation Testergebnisse
Appearance (Color) | White to Off White |
Appearance (Form) | Powder or Crystals |
Purity (HPLC area %) | ≥ 98.5 % |
Proton NMR Spectrum | Conforms to structure |
Metal trace Analysis (ICP) | Corresponds to requirements |
BARIUM (ICP) | ≤ 5 mg/kg |
CALCIUM (ICP) | ≤ 20 mg/kg |
CADMIUM (ICP) | ≤ 5 mg/kg |
COBALT (ICP) | ≤ 5 mg/kg |
CHROMIUM (ICP) | ≤ 5 mg/kg |
COPPER (ICP) | ≤ 5 mg/kg |
IRON (ICP) | ≤ 100 mg/kg |
POTASSIUM (ICP) | ≤ 20 mg/kg |
MAGNESIUM (ICP) | ≤ 5 mg/kg |
MANGANESE (ICP) | ≤ 5 mg/kg |
SODIUM (ICP) | ≤ 50 mg/kg |
NICKEL (ICP) | ≤ 5 mg/kg |
LEAD (ICP) | ≤ 5 mg/kg |
ZINC (ICP) | ≤ 5 mg/kg |
RECOMMENDED RETEST PERIOD | 48 MONTHS |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...