Organic reference standard, 2,3-Pentanedione, analytical standard, Supelco®
Lieferant: Merck
69043-100MG
69043-100MGEA 52.8 EUR
69043-100MG
Organic reference standard, 2,3-Pentanedione, analytical standard, Supelco®
Standards Organische Standards
2,3-Pentanedione can be used as a reference standard in the determination of 2,3-pentanedione in beer samples using headspace-gas chromatographic technique (HS-GC).
Verpackung: Packaged under argon.
Bitte beachten Air and light sensitive.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=71beefec0749a1218fb3c8cc3f7625d44b9f2ee7&vid=71beefec0749a1218fb3c8cc3f7625d44b9f2ee7&prid=web&clid=SAPDAM