Wasser, ULTREX® II, ultrapure, J.T.Baker®

Lieferant: Avantor

Synonyme: Wasserstoffoxid, Hydrogenoxid

6906-02EA 190.75 EUR
6906-02
Wasser, ULTREX® II, ultrapure, J.T.Baker®
Wasser
Formel: H₂O
Molecular Weight: 18.02 g/mol
Siedepunkt: 100
Schmelzpunkt: 0
Dichte: 1 g/cm³ (4 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011332
CAS-Nummer: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Bestellen

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use = 10
Color (APHA) ≤10
Chloride (Cl) ≤1 ppb
Phosphate (PO₄) ≤1 ppb
Sulfate (SO₄) ≤1 ppb
Trace Impurities - Aluminum (Al) ≤20 ppt
Trace Impurities - Antimony (Sb) ≤10 ppt
Trace Impurities - Arsenic (As) ≤10 ppt
Trace Impurities - Barium (Ba) ≤10 ppt
Trace Impurities - Beryllium (Be) ≤10 ppt
Trace Impurities - Bismuth (Bi) ≤10 ppt
Trace Impurities - Boron (B) ≤20 ppt
Trace Impurities - Cadmium (Cd) ≤10 ppt
Trace Impurities - Calcium (Ca) ≤10 ppt
Trace Impurities - Cerium (Ce) ≤10 ppt
Trace Impurities - Cesium (Cs) ≤10 ppt
Trace Impurities - Chromium (Cr) ≤10 ppt
Trace Impurities - Cobalt (Co) ≤10 ppt
Trace Impurities - Copper (Cu) ≤10 ppt
Trace Impurities - Dysprosium (Dy) ≤1 ppt
Trace Impurities - Erbium (Er) ≤1 ppt
Trace Impurities - Europium (Eu) ≤1 ppt
Trace Impurities - Gadolinium (Gd) ≤1 ppt
Trace Impurities - Gallium (Ga) ≤10 ppt
Trace Impurities - Germanium (Ge) ≤10 ppt
Trace Impurities - Gold (Au) ≤10 ppt
Trace Impurities - Hafnium (Hf) ≤1 ppt
Trace Impurities - Holmium (Ho) ≤1 ppt
Trace Impurities - Indium (In) ≤1 ppt
Trace Impurities - Iron (Fe) ≤10 ppt
Trace Impurities - Lanthanum (La) ≤1 ppt
Trace Impurities - Lead (Pb) ≤10 ppt
Trace Impurities - Lithium (Li) ≤10 ppt
Trace Impurities - Lutetium (Lu) ≤1 ppt
Trace Impurities - Magnesium (Mg) ≤10 ppt
Trace Impurities - Manganese (Mn) ≤10 ppt
Trace Impurities - Mercury (Hg) ≤20 ppt
Trace Impurities - Molybdenum (Mo) ≤10 ppt
Trace Impurities - Neodymium (Nd) ≤1 ppt
Trace Impurities - Nickel (Ni) ≤10 ppt
Trace Impurities - Niobium (Nb) ≤10 ppt
Trace Impurities - Palladium (Pd) ≤10 ppt
Trace Impurities - Platinum (Pt) ≤10 ppt
Trace Impurities - Potassium (K) ≤10 ppt
Trace Impurities - Praseodymium (Pr) ≤10 ppt
Trace Impurities - Rhenium (Re) ≤10 ppt
Trace Impurities - Rhodium (Rh) ≤10 ppt
Trace Impurities - Rubidium (Rb) ≤10 ppt
Trace Impurities - Ruthenium (Ru) ≤10 ppt
Trace Impurities - Samarium (Sm) ≤10 ppt
Trace Impurities - Scandium (Sc) ≤10 ppt
Trace Impurities - Selenium (Se) ≤50 ppt
Trace Impurities - Silver (Ag) ≤10 ppt
Trace Impurities - Sodium (Na) ≤10 ppt
Trace Impurities - Strontium (Sr) ≤10 ppt
Trace Impurities - Tantalum (Ta) ≤10 ppt
Trace Impurities - Tellurium (Te) ≤1 ppt
Trace Impurities - Terbium (Tb) ≤10 ppt
Trace Impurities - Thallium (Tl) ≤10 ppt
Trace Impurities - Thorium (Th) ≤1 ppt
Trace Impurities - Thulium (Tm) ≤10 ppt
Trace Impurities - Tin (Sn) ≤10 ppt
Trace Impurities - Titanium (Ti) ≤10 ppt
Trace Impurities - Tungsten (W) ≤10 ppt
Trace Impurities - Uranium (U) ≤1 ppt
Trace Impurities - Vanadium (V) ≤10 ppt
Trace Impurities - Ytterbium (Yb) ≤10 ppt
Trace Impurities - Yttrium (Y) ≤1 ppt
Trace Impurities - Zinc (Zn) ≤10 ppt
Trace Impurities - Zirconium (Zr) ≤10 ppt
Certificate Provided Reports Actual Lot Analysis


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR