N-Dodecyl-N,N-dimethyl-3-ammonio-1-propansulfonat (Sulfobetain-12, SB-12), Millipore®

Lieferant: Merck Millipore Calbiochem
Gefahr

Synonyme: sulphobetaine-12, 3-(Dodecyldimethylammonio)propane-1-sulphonate, N-Dodecyl-n,n-dimethyl-3-ammonio-1-propanesulphonate

693015-25 693015-5 693015-1KG
693015-25EA 682 EUR
693015-25 693015-5 693015-1KG
N-Dodecyl-N,N-dimethyl-3-ammonio-1-propansulfonat (Sulfobetain-12, SB-12), Millipore®
N-Dodecyl-N,N-dimethyl-3-ammonio-1-propansulfonat (Sulfobetain-12, SB-12)
Formel: C₁₇H₃₇NO₃S
Molecular Weight: 335.55 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00036909
CAS-Nummer: 14933-08-5
EINECS: 239-002-3
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR