Organic reference standard, 3-Methylmalic acid, ≥95,0% (HPLC), Supelco®
Lieferant: Merck
69369-10MG
69369-10MGEA 133 EUR
69369-10MG
Organic reference standard, 3-Methylmalic acid, ≥95,0% (HPLC), Supelco®
Standards Organische Standards
Metabolite of C5-branched dibasic acid metabolism, valine, leucine and isoleucine biosynthesis and 2-oxocarboxylic acid metabolism.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0fb499218d614bf5e49414973f61f08148921d41&vid=0fb499218d614bf5e49414973f61f08148921d41&prid=web&clid=SAPDAM