N-Methyl-N-trimethylsilylheptafluorobutyramide (MSHFBA) ≥90% (durch GC), LiChropur™ zur GC-Derivatisierung, Supelco®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

69484-1ML 69484-5ML
69484-1MLEA 73.4 EUR
69484-1ML 69484-5ML
N-Methyl-N-trimethylsilylheptafluorobutyramide (MSHFBA) ≥90% (durch GC), LiChropur™ zur GC-Derivatisierung, Supelco®
N-Methyl-N-trimethylsilylheptafluorobutyramide (MSHFBA)

N-methyl-N-trimethylsilylheptafluorobutyramide (MSHFBA) has been used in the silylation reaction during GC analysis of samples containing 1-monoolein, 2-monoolein, 1,2-diolein, 1,3-diolein, ethyl oleate, oleic acid and triolein.

Formel: CF₃CF₂CF₂CON(CH₃)Si(CH₃)₃
Siedepunkt: 148 °C (Lit.)
Dichte: 1,254 g/mL (25 °C) (Lit.)
Flammpunkt: 33 °C
MDL: MFCD00077822
CAS-Nummer: 53296-64-3
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a5893fbdcbd45aece52d605a4faf060ce574b19a&vid=eacf81a3229667f578b181a1d23d67c02bbbe0d8&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) Colorless to Very Light Yellow
Appearance (Form) Liquid
Purity (GC area %) ≥ 90.0 %
Refractive index n20/D 1.351 - 1.355
Infrared spectrum Conforms to structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR