Morin Hydrat 1-2 mol/mol water for die Bestimmung von Al, Be, Zn, Ga, In, Sc, für die Mikroskopie, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: 2',3,4',5,7-Pentahydroxyflavone

M-Clarity™ Qualitätsstufe = MQ200

69870-10G 69870-50G 69870-5G
69870-10GEA 151 EUR
69870-10G 69870-50G 69870-5G
Morin Hydrat 1-2 mol/mol water for die Bestimmung von Al, Be, Zn, Ga, In, Sc, für die Mikroskopie, Sigma-Aldrich®
Morin Hydrat

Morin hydrate is a yellowish pigment and a fluorescent indicator. It is used for the fluorimetric determination of several metals, mostly aluminum. Morin has been utilised in fluorescent microscopy as a fluorescent nuclear stain and as an application to detect aluminium in the brain by demonstrating metal salts.

Formel: C₁₅H₁₀O₇
Molecular Weight: 302.24 g/mol
Schmelzpunkt: 299...300 °C
Lagertemperatur: Room Temperature
MDL: MFCD00217054
CAS-Nummer: 654055-01-3
EINECS: 207-542-9

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) Faint Brown to Brown and Faint Brown-Yellow to Brown Yellow
Appearance (Form) Powder
Purity (HPLC) ≥85%
Thin Layer Chromatography Corresponds to Standard
Solubility (Colour) Brown to Very Dark Brown and Red to Very Dark Red
Water 3 - 11 %
Sulphated Ash ≤3%
Infrared Spectrum Conforms to Structure
Wavelength (1) (UV) 360 - 375 nm
Extinction (1) (UV) (1%/1 cm) ≥300
Solvent (UV) Methanol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR