Molybdate reagent solution, for photometric phosphate determination acc. to DIN 38405-D11-1, Supelco®
Lieferant: Merck
69888-100ML
69888-100MLEA 38.9 EUR
69888-100ML
Molybdate reagent solution, for photometric phosphate determination acc. to DIN 38405-D11-1, Supelco®
Umwelt-Prüfkits
Solution contains hexaammonium heptamolybdate ((NH₄)₆Mo₇O₂₄.4H₂O); sulfuric acid (H₂SO₄); potassium antimony(III) oxide tartrate(K(SbO)C₄H₄O₆.0.5H₂O);water
Bitte beachten Solution is corrosive (GHS05) please consult MSDS.