Top Works™-Systeme für Medienflaschen-Verschlüsse, Nalgene®
Lieferant: Thermo Fisher Scientific
2132-1003
700-0001EA 380 EUR
700-0001
Top Works™-Systeme für Medienflaschen-Verschlüsse, Nalgene®
Verschlüsse Flaschenverschlüsse
PSF, closures with platinum cured silicone inserts.
  • They fit the corresponding threads on bottles by Schott, Corning and Wheaton
  • For thread size GL 45
  • Autoclavable, leakproof
These closures make it easier to transfer liquids aseptically from most conventional glass media bottles.