Blotting papers, Owl™
Lieferant: THERMO OWL SCIENTIFIC
FP-1 FP-2 FP-3 FP-4 FP-5 FP-6 FP-7
700-2190EA 159 EUR
700-2190 OWLAFP-2 OWLAFP-3 700-2191 700-2192 OWLAFP-6 OWLAFP-7
Blotting papers, Owl™
Blotting-Papiere
Absorb adequate amounts of buffer for semi-dry electroblotting without the loss of mechanical stability with Thermo Scientific™ Owl™ Blotting Paper, high-quality blotting paper ideal for wicking and blotting applications.
Bestellen
TECHNISCHE DATEN
Environmentally Preferable
Gewicht 183 g/m² 183 g/m²
Stärke 0,83 mm 0,83 mm