3,3'-Diethyloxacarbocyanine iodide [DiOC2(3) iodide] Membranfarbstoff, für die Histologie

Lieferant: Biotium

Synonyme: DOC Iodide, 3-Ethyl-2-[3-[3-ethyl-2(3H)-benzoxazolylidene]-1-propenyl]benzoxazolium Iodide, 3,3'-Diethyloxacarbocyanine iodide, DiOC2(3) iodide, 3-Ethyl-2-(3-(3-ethylbenzo[d]oxazol-2(3H)-ylidene)prop-1-en-1-yl)benzo[d]oxazol-3-ium iodide

70008
70008.EA 321 EUR
70008.
3,3'-Diethyloxacarbocyanine iodide [DiOC2(3) iodide] Membranfarbstoff, für die Histologie
3,3'-Diethyloxacarbocyanine iodide [DiOC2(3) iodide]

DiOC2(3) (3,3′-Diethyloxacarbocyanine, iodide) has been used for measuring membrane potentials in bacteria. The green fluorescent dye forms red fluorescent aggregates with increasing membrane potential. The spectral shifting phenomenon has been exploited to measure membrane potential ratiometrically.


  • Ex/Em (MeOH) = 482/497 nm
  • Orange solid soluble in DMSO or DMF
  • Store at 4°C and protect from light

Formel: C₂₁H₂₁IN₂O₂
Molecular Weight: 460,32 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00011952
CAS-Nummer: 905-96-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3181900aa224607aed31fd79980b6d9ecaad87da&vid=3181900aa224607aed31fd79980b6d9ecaad87da&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR