3,3'-Di-n-hexyloxacarbocyanine iodide [DiOC6(3) iodide] Membranfarbstoff, für die Histologie

Lieferant: Biotium
Achtung

Synonyme: 3-Hexyl-2-[3-(3-hexyl-1,3-benzoxazol-2(3H)-ylidene)-1-propen-1-yl]-1,3-benzoxazol-3-ium iodide, DiOC6(3) iodide, DiOC6(3), 3,3'-Dihexyloxacarbocyanine iodide, 3-Hexyl-2-[3-(3-hexyl-2(3H)benzoxazolylidene)-1-propenyl]benzoxazolium iodide

70009
70009.EA 307 EUR
70009.
3,3'-Di-n-hexyloxacarbocyanine iodide [DiOC6(3) iodide] Membranfarbstoff, für die Histologie
3,3'-Di-n-hexyloxacarbocyanine iodide [DiOC6(3) iodide]

DiOC6(3) (3,3′-Dihexyloxacarbocyanine,iodide) is a green fluorescent membrane dye that has been used to stain the ER in both live and fixed cells. The dye has also been used to study structural interactions and dynamics of the ER in neurons and yeast. In addition, the dye is widely used as a membrane potential dye.


  • λEx/λEm (MeOH) = 484/501 nm
  • Orange solid soluble in DMSO or DMF
  • Store at 4°C and protect from light, especially in solution

Formel: C₂₉H₃₇IN₂O₂
Molecular Weight: 572.531 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00011954
CAS-Nummer: 53213-82-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b4ef09dc629108b0a0b7010f6651f8beddf32f8a&vid=b4ef09dc629108b0a0b7010f6651f8beddf32f8a&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR