Acridine Orange 10-nonyl bromide (NAO) Mitochondrien-Färbemittel, für die Fluoreszenzmikroskopie
Lieferant: Biotium
|
Achtung
|
Synonyme:
3,6-Bis(dimethylamino)-10-nonylacridinium bromide, Nonyl acridine orange, NAO, 10-Nonylacridine orange bromide
NAO is a green fluorescent mitochondrial dye whose staining is not dependent on mitochondrial membrane potential.
- Cellular localization: Mitochondria
- For live or fixed cells: For live/intact cells
- Assay type/options: No-wash staining, Real-time imaging
- Potential dependence: Mitochondrial potential-independent
- Colors: Red
- Excitation/Emission: 495/522 nm
- Orange solid soluble in DMSO or DMF
- Store at 4°C and protect from light, especially in solution
NAO is a green fluorescent mitochondrial dye whose staining is not dependent on mitochondrial membrane potential, unlike JC-1 (70011) or Rhodamine 123 (70010). NAO has been used to study multidrug resistance and to measure changes in mitochondrial mass during apoptosis. The dye is reported to bind cardiolipin, a phosphoprotein in the inner mitochondrial membrane. Also see our MitoView™ Dyes and other mitochondrial dyes.
Formel:
C₂₆H₃₈BrN₃ Molecular Weight: 472.50 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00467605 CAS-Nummer: 75168-11-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...