DiIC1(5) iodide Mitochondrien-Färbemittel, für die Fluoreszenzmikroskopie
Lieferant: Biotium
Synonyme:
1,1’,3,3,3’,3’-Hexamethylindodicarbocyanine iodide
70015
70015.EA
270
EUR
70015.
DiIC1(5) iodide Mitochondrien-Färbemittel, für die Fluoreszenzmikroskopie
DiIC1(5) iodide
The carbocyanine dye DiIC1(5) has been used to measure mitochondrial membrane potential in apoptotic cells. Loss of membrane potential in apoptotic cells is reflected by a decrease of fluorescence signal in the infrared channel.
- Cellular localization: Mitochondria
- For live or fixed cells: For live/intact cells
- Assay type/options: Real-time imaging
- Cell permeability: Membrane permeant
- Apoptosis/viability marker: Mitochondrial potential
- Potential dependence: Mitochondrial potential-dependent
- Colors: Far-red
- Excitation/Emission: 638/658 nm
- Dark green solid soluble in DMSO or DMF
- Store at 4°C and protect from light
Formel:
C₂₇H₃₁IN₂ Molecular Weight: 510,5 g/mol Lagertemperatur: Kühlschrank |
CAS-Nummer:
36536-22-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...