Tetramethylrhodamine ethyl ester perchlorate (TMRE) ≥95% (durch HPLC) Mitochondrien-Färbemittel, geeignet für Fluoreszenz
Lieferant: Biotium
|
Achtung
|
Synonyme:
Tetramethylrhodamine Ethyl Ester Perchlorate, TMRE, 3,6-bis(Dimethylamino)-9-[2-(ethoxycarbonyl)phenyl]-xanthylium perchlorate
TMRE (Tetramethylrhodamine ethyl ester, perchlorate) and TMRM (70017) are preferred dyes for quantitative measurements of membrane potentials using the Nernst equation.
- Cellular localization: Mitochondria
- For live or fixed cells: For live/intact cells
- Assay type/options: Real-time imaging
- Cell permeability: Membrane permeant
- Apoptosis/viability marker: Mitochondrial potential
- Potential dependence: Mitochondrial potential-dependent
- λEx/λEm (MeOH) = 549/574 nm
- Red solid soluble in DMSO, DMF or EtOH
- Store at 4°C and protect from light
TMRE (Tetramethylrhodamine ethyl ester, perchlorate) and TMRM (70017) are preferred dyes for quantitative measurements of membrane potentials using the Nernst equation. The dyes do not form aggregates in cell membranes and have minimal interaction with membrane proteins. Thus, the transmembrane distribution of the dyes is directly related to the membrane potential according to the Nernst equation. Biotium also offers TMRE, 2 mM in DMSO (70005) as a ready to use stock solution.
Formel:
C₂₆H₂₇ClN₂O₇ Molecular Weight: 514,95 g/mol |
MDL:
MFCD00467973 CAS-Nummer: 115532-52-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...